Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039803.2 | NP_001034892.1 | 2 | 510 | ||
NM_001170639.1 | NP_001164110.1 | 2 | 510 | |||
NM_001170640.1 | NP_001164111.1 | 2 | 510 | |||
NM_012119.4 | NP_036251.2 | 2 | 510 | |||
NM_178432.3 | NP_848519.1 | 2 | 549 | |||
XM_017014561.1 | XP_016870050.1 | 2 | 440 | |||
XM_017014562.1 | XP_016870051.1 | 2 | 440 | |||
XM_017014563.1 | XP_016870052.1 | 2 | 440 | |||
XM_017014564.1 | XP_016870053.1 | 2 | 440 | |||
GenBank mRNA | AF035013.1 | - | 2 | 413 | ||
AF113130.1 | - | 2 | 421 | |||
AK075325.1 | - | 2 | 302 | |||
AK226135.1 | - | 1 | 600 | |||
AK291713.1 | - | 2 | 431 | |||
AK294193.1 | - | 2 | 263 | |||
AK296328.1 | - | 2 | 261 | |||
AK298993.1 | - | 2 | 253 | |||
AY904367.1 | - | 2 | 176 | |||
BC002655.2 | - | 2 | 235 | 3605918 | ||
BF966262.1 | - | 2 | 488 | |||
BI757318.1 | - | 2 | 361 | |||
BX647489.1 | - | 2 | 439 | |||
CB992644.1 | - | 2 | 519 | |||
KU178572.1 | - | 2 | 176 | |||
KU178573.1 | - | 2 | 176 | |||
KU178574.1 | - | 2 | 176 | |||
KU178575.1 | - | 2 | 176 | |||
KU178576.1 | - | 2 | 176 | |||
KU178577.1 | - | 2 | 176 |