Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134012.3 | NP_598773.2 | 8, 9 | 1089 | ||
GenBank mRNA | AK028503.1 | - | 1, 2 | 31 | ||
AK033238.1 | - | 3, 4 | 3962 | |||
AK141155.1 | - | 8, 9 | 1084 | |||
AK153919.1 | - | 10, 11 | 924 | |||
BC020018.1 | - | 4, 5 | 400 | 2646754 | ||
BC062907.1 | - | 8, 9 | 1089 | 6813849 | ||
BC064014.1 | - | 10, 11 | 1193 | 6335954 |