Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035226.1 | NP_001030303.1 | 17 | 1951 | ||
NM_134014.3 | NP_598775.2 | 17 | 2189 | |||
GenBank mRNA | AK136538.1 | - | 10 | 1352 | ||
AK142055.1 | - | 1 | 113 | |||
BC012276.1 | - | 4 | 413 | 3156478 | ||
BC025628.1 | - | 3 | 373 | 5321933 | ||
BC029714.1 | - | 15 | 1712 | |||
BC034710.1 | - | 16 | 1946 | |||
BC062912.1 | - | 17 | 1951 | 6821729 | ||
BU052960.1 | - | 3 | 294 |