Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134024.2 | NP_598785.1 | 4 | 419 | ||
GenBank mRNA | AB158480.1 | - | 4 | 431 | ||
AK154019.1 | - | 4 | 387 | |||
AK160125.1 | - | 4 | 433 | |||
BC006581.1 | - | 4 | 419 | 3587066 |