Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134025.3 | NP_598786.1 | 3 | 588 | ||
GenBank mRNA | AK035552.1 | - | 3 | 589 | ||
AK041175.1 | - | 3 | 684 | |||
AK153798.1 | - | 2 | 795 | |||
AK166413.1 | - | 3 | 632 | |||
BC021800.1 | - | 2 | 412 | 3671142 |