Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 4 | 711 | ||
GenBank mRNA | AK037253.1 | - | 4 | 643 | ||
AK041513.1 | - | 4 | 656 | |||
AK044742.1 | - | 4 | 653 | |||
AK090014.1 | - | 4 | 687 | |||
AK129344.1 | - | 4 | 622 | |||
AK132660.1 | - | 4 | 233 | |||
AK165940.1 | - | 4 | 635 | |||
AK166627.1 | - | 4 | 637 | |||
BC006870.1 | - | 4 | 622 | 3595439 |