Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134050.4 | NP_598811.3 | 3 | 421 | ||
GenBank mRNA | AB232610.1 | - | 3 | 214 | ||
AK158637.1 | - | 3 | 343 | |||
AK158768.1 | - | 3 | 375 | |||
BC013790.1 | - | 3 | 367 | 2655151 | ||
BC027769.1 | - | 3 | 421 | 5345297 |