Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 9 | 1799 | ||
GenBank mRNA | AK003405.1 | - | 745 | |||
AK078906.1 | - | 9 | 2120 | |||
BC022707.1 | - | 7 | 1226 | 4486171 | ||
BC026630.1 | - | 9 | 1799 | 4983657 | ||
BC032195.1 | - | 10 | 1663 | 5323331 |