Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 2 | 470 | ||
NM_001010931.2 | NP_001010931.1 | 2 | 470 | |||
NM_001010932.2 | NP_001010932.1 | 2 | 470 | |||
NM_001010933.2 | NP_001010933.1 | 2 | 470 | |||
NM_001010934.2 | NP_001010934.1 | 2 | 470 | |||
XM_006715956.2 | XP_006716019.1 | 3 | 433 | |||
XM_011516115.2 | XP_011514417.1 | 3 | 433 | |||
XM_017012097.1 | XP_016867586.1 | 3 | 433 | |||
XM_017012098.1 | XP_016867587.1 | 3 | 433 | |||
GenBank mRNA | AB208900.1 | - | 2 | 377 | ||
AK291674.1 | - | 2 | 405 | |||
AK313694.1 | - | 2 | 320 | |||
BC022308.1 | - | 2 | 319 | |||
BC063485.1 | - | 2 | 328 | |||
BC105797.1 | - | 2 | 410 | |||
BC130284.1 | - | 2 | 318 | 40146315 | ||
BC130286.1 | - | 2 | 318 | 40146317 | ||
BU533235.1 | - | 409 | ||||
FJ830861.1 | - | 2 | 243 | |||
FJ830862.1 | - | 2 | 243 | |||
L02931.1 | - | 2 | 303 | |||
M29145.1 | - | 2 | 344 | |||
M55379.1 | - | 2 | 243 | |||
M60718.1 | - | 2 | 315 | |||
M73239.1 | - | 2 | 308 | |||
M73240.1 | - | 2 | 308 | |||
M77227.1 | - | 2 | 325 | |||
U46010.1 | - | 2 | 297 | |||
X57574.1 | - | 2 | 320 |