Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133947.3 | NP_598708.3 | 12, 13 | 1271 | ||
GenBank mRNA | AK032693.1 | - | 12, 13 | 1320 | ||
AK050670.1 | - | 12, 13 | 1353 | |||
AK168399.1 | - | 12, 13 | 1289 | |||
BC049791.1 | - | 12, 13 | 1271 | 6505037 |