Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281974.1 | NP_001268903.1 | 7 | 1969 | ||
NM_001281975.1 | NP_001268904.1 | 7 | 2019 | |||
NM_133971.3 | NP_598732.2 | 6 | 1857 | |||
GenBank mRNA | AK081754.1 | - | 7 | 2017 | ||
AK082875.1 | - | 6 | 1856 | |||
AK084456.1 | - | 7 | 1967 | |||
AK086012.1 | - | 4 | 2222 | |||
AK088371.1 | - | 6 | 1807 | |||
AK200034.1 | - | 1 | 116 | |||
BC002198.1 | - | 6 | 1789 | 3489133 |