Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080147.1 | - | 5 | 652 | ||
AK082889.1 | - | 5 | 647 | |||
AK089984.1 | - | 5 | 683 | |||
AK143292.1 | - | 5 | 652 | |||
AK154675.1 | - | 5 | 707 | |||
BC021472.1 | - | 1 | 106 | |||
BC052425.1 | - | 5 | 675 | 6835500 | ||
BC059939.1 | - | 5 | 643 | 6477224 | ||
S80963.1 | - | 5 | 680 |