Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134034.2 | NP_598795.2 | 4 | 1186 | ||
GenBank mRNA | AK037253.1 | - | 4 | 1118 | ||
AK041513.1 | - | 4 | 1131 | |||
AK044742.1 | - | 4 | 1129 | |||
AK129344.1 | - | 4 | 1097 | |||
AK132660.1 | - | 4 | 708 | |||
AK165940.1 | - | 4 | 1111 | |||
AK166627.1 | - | 4 | 1112 | |||
BC006870.1 | - | 4 | 1097 | 3595439 |