Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253847.1 | NP_001240776.1 | 6 | 1641 | ||
NM_001253848.1 | NP_001240777.1 | 7 | 1164 | |||
NM_134048.2 | NP_598809.1 | 6 | 1638 | |||
GenBank mRNA | AF167441.1 | - | 6 | 1641 | ||
AK037706.1 | - | 3 | 1225 | |||
AK049141.1 | - | 8 | 1349 | |||
AK051737.1 | - | 6 | 1580 | |||
AK166017.1 | - | 7 | 1108 | |||
BC019529.1 | - | 6 | 1562 | 4219836 |