Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134091.2 | NP_598852.2 | 3 | 267 | ||
GenBank mRNA | AK142314.1 | - | 3 | 234 | ||
AK164757.1 | - | 3 | 196 | |||
AK166391.1 | - | 2 | 222 | |||
AK170806.1 | - | 3 | 234 | |||
AY382616.1 | - | 3 | 221 | |||
BC018197.1 | - | 3 | 227 | 4190407 | ||
BC024122.1 | - | 3 | 227 | 5097970 | ||
BC025561.1 | - | 3 | 205 | 5322980 | ||
BY242851.1 | - | 3 | 269 |