Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134115.2 | NP_598876.1 | 3 | 387 | ||
GenBank mRNA | AK035731.1 | - | 3 | 396 | ||
AK080179.1 | - | 3 | 396 | |||
AK084433.1 | - | 3 | 386 | |||
AK184259.1 | - | 4 | 291 | |||
AY292399.1 | - | 3 | 251 | |||
BC009658.1 | - | 3 | 387 | 3484764 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK153676.1 | - | 332 | Chr. - - [-] on Build |