Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134133.2 | NP_598894.1 | 2 | 876 | ||
GenBank mRNA | AF313412.1 | - | 2 | 418 | ||
AK143383.1 | - | 2 | 880 | |||
AK155873.1 | - | 2 | 522 | |||
AK171810.1 | - | 2 | 534 | |||
BC012898.1 | - | 2 | 384 | 4013357 |