Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134135.1 | NP_598896.1 | 1 | 183 | ||
GenBank mRNA | AK087519.1 | - | 3 | 340 | ||
BC005502.1 | - | 1 | 183 | 3490154 | ||
BC085175.1 | - | 3 | 323 |