Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 2 | 192 | ||
GenBank mRNA | AK076116.1 | - | 2 | 148 | ||
AK079615.1 | - | 2 | 154 | |||
AK088757.1 | - | 1 | 119 | |||
AK136821.1 | - | 2 | 186 | |||
AK146282.1 | - | 2 | 140 | |||
AK163705.1 | - | 2 | 171 | |||
AK168252.1 | - | 2 | 142 | |||
AK181039.1 | - | 1 | 82 | |||
AK186862.1 | - | 2 | 192 | |||
AK189220.1 | - | 1 | 83 | |||
AK220040.1 | - | 1 | 82 | |||
AY523555.1 | - | 1 | 113 | |||
BC016261.1 | - | 1 | 110 | 4920560 | ||
BC021864.1 | - | 2 | 132 | 5123958 | ||
BY283649.1 | - | 2 | 211 |