Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134077.4 | NP_598838.3 | 11 | 2041 | ||
GenBank mRNA | AK049083.1 | - | 10 | 1496 | ||
AK051686.1 | - | 11 | 2017 | |||
AK139077.1 | - | 11 | 2042 | |||
AK172039.1 | - | 11 | 2020 | |||
BC049360.1 | - | 11 | 2018 | |||
BC066051.1 | - | 11 | 1993 |