Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134093.2 | NP_598854.1 | 2, 3 | 372 | ||
GenBank mRNA | AF287293.1 | - | 2, 3 | 312 | ||
AF401483.1 | - | 2, 3 | 205 | |||
AK041312.1 | - | 1, 2 | 701 | |||
AK045256.1 | - | 2, 3 | 192 | |||
AK080221.1 | - | 2, 3 | 292 | |||
AK154676.1 | - | 2, 3 | 287 | |||
AK169341.1 | - | 2, 3 | 292 | |||
BY293457.1 | - | 2, 3 | 372 |