Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 2 | 243 | ||
GenBank mRNA | AK076116.1 | - | 2 | 199 | ||
AK079615.1 | - | 2 | 205 | |||
AK088757.1 | - | 1 | 170 | |||
AK136821.1 | - | 2 | 237 | |||
AK146282.1 | - | 2 | 191 | |||
AK163705.1 | - | 2 | 222 | |||
AK168252.1 | - | 2 | 193 | |||
AK186862.1 | - | 2 | 243 | |||
AK200007.1 | - | 1 | 132 | |||
AY523555.1 | - | 1 | 164 | |||
BC016261.1 | - | 1 | 161 | 4920560 | ||
BC021864.1 | - | 2 | 183 | 5123958 | ||
BY283649.1 | - | 2 | 262 |