Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134072.1 | NP_598833.1 | 7, 8 | 895 | ||
GenBank mRNA | AK033537.1 | - | 7, 8 | 913 | ||
AK136528.1 | - | 7, 8 | 912 | |||
AY730283.1 | - | 7, 8 | 857 | |||
BC013482.1 | - | 7, 8 | 895 | 4240772 | ||
BC026628.1 | - | 7, 8 | 894 | 4983237 |