Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 6 | 1575 | ||
NM_001282543.1 | NP_001269472.1 | 5 | 1518 | |||
NR_104212.1 | 5 | 1426 | ||||
NR_104215.1 | 4 | 1369 | ||||
NR_104216.1 | 5 | 625 | ||||
XM_017004613.1 | XP_016860102.1 | 7 | 1623 | |||
XM_017004614.1 | XP_016860103.1 | 7 | 1623 | |||
GenBank mRNA | AK223409.1 | - | 6 | 1569 | ||
AK301843.1 | - | 5 | 1388 | |||
AK314260.1 | - | 6 | 1535 | |||
AX794378.1 | - | 6 | 1506 | |||
BC126426.1 | - | 6 | 1503 | 8992142 | ||
BC126428.1 | - | 6 | 1503 | 8992144 | ||
JF790280.1 | - | 5 | 1449 | |||
JF790281.1 | - | 4 | 1300 | |||
JF790287.1 | - | 6 | 1053 | |||
U76638.1 | - | 6 | 1506 |