Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134081.5 | NP_598842.1 | 5 | 903 | ||
GenBank mRNA | AK075979.1 | - | 5 | 864 | ||
AK077324.1 | - | 5 | 873 | |||
AK153033.1 | - | 5 | 863 | |||
AK162075.1 | - | 5 | 778 | |||
BC014686.1 | - | 5 | 865 | 4481661 | ||
BC023787.1 | - | 5 | 861 | 5346759 | ||
BC027012.1 | - | 5 | 856 | 5367851 |