Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 10, 11 | 1212 | ||
NM_134083.4 | NP_598844.3 | 10, 11 | 1207 | |||
GenBank mRNA | AK013540.1 | - | 1317 | |||
AK031242.1 | - | 10, 11 | 1173 | |||
AK031248.1 | - | 10, 11 | 1178 | |||
AK147784.1 | - | 11, 12 | 1213 | |||
AK160881.1 | - | 11, 12 | 1399 | |||
BC003224.1 | - | 8, 9 | 1005 | 3482729 | ||
JN086338.1 | - | 9, 10 | 1128 | |||
JN086339.1 | - | 10, 11 | 1366 |