Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 14 | 1555 | ||
NM_001166458.1 | NP_001159930.1 | 14 | 1762 | |||
NM_134086.4 | NP_598847.2 | 14 | 2248 | |||
GenBank mRNA | AF184240.2 | - | 13 | 1340 | ||
AK034130.1 | - | 14 | 2250 | |||
AK035098.1 | - | 13 | 1267 | |||
AK047459.1 | - | 14 | 1779 | |||
AK049173.1 | - | 14 | 1464 | |||
AK049294.1 | - | 14 | 1528 | |||
AK081724.1 | - | 14 | 1764 | |||
AK082683.1 | - | 14 | 1466 | |||
AK162612.1 | - | 14 | 1613 | |||
AK163914.1 | - | 9 | 866 | |||
AK165188.1 | - | 14 | 1815 | |||
AK204713.1 | - | 2 | 261 | |||
BC030378.1 | - | 14 | 1464 | 5375319 |