Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134092.3 | NP_598853.3 | 17 | 2134 | ||
GenBank mRNA | AJ278508.1 | - | 17 | 2055 | ||
AK080036.1 | - | 17 | 1948 | |||
AK088652.1 | - | 7 | 1562 | |||
AK133855.1 | - | 17 | 1982 | |||
BC005500.1 | - | 10 | 1131 | |||
BC052197.1 | - | 17 | 2134 | 6511752 |