Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113401.1 | NP_001106872.1 | 6 | 894 | ||
NM_001113405.1 | NP_001106876.1 | 4 | 536 | |||
NM_134111.2 | NP_598872.1 | 6 | 884 | |||
GenBank mRNA | AB081298.1 | - | 4 | 536 | ||
AK076871.1 | - | 7 | 831 | |||
AY034479.1 | - | 6 | 901 | |||
AY049021.1 | - | 6 | 840 | |||
BC004721.1 | - | 6 | 883 | 3582359 | ||
BC056626.1 | - | 6 | 884 | 5008372 |