Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253843.1 | NP_001240772.1 | 14, 15 | 1595 | ||
NM_001253844.1 | NP_001240773.1 | 12, 13 | 1188 | |||
NM_134129.4 | NP_598890.1 | 14, 15 | 1538 | |||
GenBank mRNA | AB222602.1 | - | 14, 15 | 1380 | ||
AF251503.1 | - | 14, 15 | 1503 | |||
AK047693.1 | - | 14, 15 | 1413 | |||
AK076998.1 | - | 12, 13 | 1189 | |||
AK088769.1 | - | 14, 15 | 1523 | |||
AK164681.1 | - | 14, 15 | 1526 | |||
AK183921.1 | - | 2, 3 | 273 | |||
AK196388.1 | - | 2, 3 | 273 | |||
AK208164.1 | - | 2, 3 | 273 | |||
AK210572.1 | - | 4, 5 | 380 | |||
BC004070.1 | - | 14, 15 | 1477 | 3591050 |