Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134133.2 | NP_598894.1 | 2 | 1038 | ||
GenBank mRNA | AF313412.1 | - | 2 | 580 | ||
AK143383.1 | - | 2 | 1042 | |||
AK155873.1 | - | 2 | 684 | |||
AK168283.1 | - | 2 | 861 | |||
AK171810.1 | - | 2 | 696 | |||
BC012898.1 | - | 2 | 546 | 4013357 |