Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134139.1 | NP_598900.1 | 9, 10 | 932 | ||
GenBank mRNA | AK077521.1 | - | 9, 10 | 933 | ||
AK145704.1 | - | 9, 10 | 813 | |||
BC019968.1 | - | 9, 10 | 932 | 4167260 | ||
BC024478.1 | - | 5, 6 | 527 | 4977054 |