Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 3 | 294 | ||
GenBank mRNA | AK076116.1 | - | 3 | 250 | ||
AK079615.1 | - | 3 | 256 | |||
AK088757.1 | - | 2 | 221 | |||
AK136821.1 | - | 3 | 288 | |||
AK146282.1 | - | 3 | 242 | |||
AK163705.1 | - | 3 | 273 | |||
AK168252.1 | - | 3 | 244 | |||
AK181039.1 | - | 2 | 150 | |||
AK188252.1 | - | 2 | 39 | |||
AK192608.1 | - | 1 | 39 | |||
AK210999.1 | - | 2 | 39 | |||
AK220040.1 | - | 2 | 150 | |||
AY523555.1 | - | 2 | 215 | |||
BC016261.1 | - | 2 | 212 | 4920560 | ||
BC021864.1 | - | 3 | 234 | 5123958 | ||
BC021949.1 | - | 2 | 45 | |||
BY283649.1 | - | 3 | 313 |