Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134142.1 | NP_598903.1 | 4 | 1829 | ||
GenBank mRNA | AK003502.1 | - | 1 | 314 | ||
AK049148.1 | - | 4 | 1840 | |||
AK150779.1 | - | 4 | 1813 | |||
AK151850.1 | - | 4 | 1810 | |||
AK152419.1 | - | 4 | 1810 | |||
AK165990.1 | - | 4 | 1787 | |||
AK196648.1 | - | 1 | 290 | |||
BC022997.1 | - | 4 | 1829 | 5362560 | ||
BC023901.1 | - | 4 | 1785 | 5321274 | ||
BC025033.1 | - | 4 | 1797 | 4947460 | ||
BC025815.1 | - | 4 | 1819 | 5097938 |