Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134152.3 | NP_598913.1 | 8 | 846 | ||
GenBank mRNA | AB053936.2 | - | 8 | 898 | ||
AB071194.1 | - | 8 | 871 | |||
AK077085.1 | - | 8 | 882 | |||
BC026563.1 | - | 8 | 812 | 5065405 |