Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000485.2 | NP_000476.1 | 5 | 680 | ||
NM_001030018.1 | NP_001025189.1 | 5 | 546 | |||
GenBank mRNA | BM423481.1 | - | 704 | |||
BM550173.1 | - | 5 | 569 | |||
BU507629.1 | - | 1 | 81 | |||
CR749423.1 | - | 5 | 671 |