Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134420.4 | NP_599252.2 | 1292 | |||
GenBank mRNA | AB099881.1 | - | 453 | |||
AF248494.1 | - | 638 | ||||
AK029238.1 | - | 2218 | ||||
AK046993.1 | - | 2436 | ||||
AK053088.1 | - | 827 | ||||
AK080750.1 | - | 651 | ||||
AK163670.1 | - | 1293 | ||||
AK163671.1 | - | 1293 | ||||
AY032863.1 | - | 597 | ||||
AY049076.1 | - | 552 | ||||
BC028856.1 | - | 616 | 4165725 |