Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199349.1 | NP_001186278.1 | 1, 2 | 336 | ||
NM_138309.3 | NP_612182.1 | 1, 2 | 336 | |||
GenBank mRNA | AK031264.1 | - | 1, 2 | 93 | ||
AK054364.1 | - | 1, 2 | 136 | |||
AK080225.1 | - | 1, 2 | 92 | |||
AK166275.1 | - | 1, 2 | 87 | |||
AY078163.1 | - | 1, 2 | 132 | |||
BC031736.1 | - | 1, 2 | 111 | 4013657 | ||
BY727540.1 | - | 1, 2 | 337 | |||
EF516990.1 | - | 1, 2 | 94 |