Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045863.1 | NP_001039328.1 | 3 | 295 | ||
NM_001045864.1 | NP_001039329.1 | 3 | 381 | |||
NM_138580.2 | NP_613046.2 | 3 | 169 | |||
GenBank mRNA | AK011663.1 | - | 3 | 153 | ||
AK012389.1 | - | 3 | 186 | |||
AK131947.1 | - | 3 | 153 | |||
AK147011.1 | - | 3 | 277 | |||
BC025905.1 | - | 1 | 233 | |||
BC034867.1 | - | 3 | 121 | 1495547 | ||
CN525174.1 | - | 3 | 124 |