Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700088E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138581.2 | NP_613047.1 | 3 | 228 | ||
GenBank mRNA | AA008475.1 | - | 3 | 217 | ||
AK006539.1 | - | 3 | 209 | |||
AK030004.1 | - | 314 | ||||
BC068203.1 | - | 2 | 273 | |||
BC096672.1 | - | 2 | 326 | 5117646 | ||
BC116891.1 | - | 3 | 159 | 40126210 | ||
BC116893.1 | - | 3 | 159 | 40126212 | ||
BC144714.1 | - | 3 | 159 | 9053262 | ||
BC144715.1 | - | 3 | 271 |