Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138603.3 | NP_613069.3 | 3, 4 | 496 | ||
GenBank mRNA | AF229637.1 | - | 3, 4 | 437 | ||
AK010536.1 | - | 3, 4 | 414 | |||
AK039573.1 | - | 3, 4 | 469 | |||
AK209776.1 | - | 2, 3 | 174 | |||
BC011195.1 | - | 3, 4 | 401 | 4188459 |