Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138646.3 | NP_619587.3 | 3 | 619 | ||
GenBank mRNA | AK081489.1 | - | 3 | 579 | ||
AK129418.1 | - | 2 | 506 | |||
AK147954.1 | - | 2 | 1062 | |||
AK171871.1 | - | 3 | 588 | |||
AY043415.1 | - | 2 | 139 | |||
BC063055.1 | - | 2 | 311 | 5701206 | ||
BY741149.1 | - | 3 | 619 |