Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102468.1 | NP_001095938.1 | 3 | 162 | ||
NM_138304.2 | NP_612177.1 | 4 | 303 | |||
GenBank mRNA | AK008039.1 | - | 4 | 289 | ||
AK146656.1 | - | 4 | 276 | |||
AY061807.1 | - | 4 | 303 | |||
BC013541.1 | - | 4 | 281 | 4241711 | ||
BC023475.1 | - | 4 | 268 | 5042406 | ||
BY097052.1 | - | 3 | 175 |