Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138310.1 | NP_612183.1 | 2 | 198 | ||
GenBank mRNA | AF141335.1 | - | 2 | 198 | ||
AK156422.1 | - | 2 | 265 | |||
BC030718.1 | - | 2 | 206 | 4482457 |