Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138586.3 | NP_613052.1 | 2 | 280 | ||
GenBank mRNA | AK006475.1 | - | 2 | 233 | ||
AK007372.1 | - | 2 | 269 | |||
AK137780.1 | - | 243 | ||||
AK155228.1 | - | 2 | 232 | |||
BB856072.1 | - | 2 | 282 | |||
BC034358.1 | - | 2 | 226 | 2651418 | ||
BC092393.1 | - | 2 | 221 | |||
BY196593.1 | - | 2 | 278 | |||
BY198385.1 | - | 2 | 278 |