Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138591.2 | NP_613057.2 | 2, 3 | 369 | ||
GenBank mRNA | AF315511.1 | - | 2, 3 | 336 | ||
AK033214.1 | - | 2, 3 | 368 | |||
BC013093.1 | - | 2, 3 | 372 | 3584730 | ||
BC031772.1 | - | 2, 3 | 336 | 4239980 |