Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138592.4 | NP_613058.1 | 5, 6 | 754 | ||
GenBank mRNA | AK010443.1 | - | 5, 6 | 764 | ||
AK018595.1 | - | 5, 6 | 755 | |||
AK033105.1 | - | 5, 6 | 751 | |||
AK045062.1 | - | 5, 6 | 751 | |||
AK088354.1 | - | 5, 6 | 766 | |||
AK165625.1 | - | 5, 6 | 735 | |||
AK167668.1 | - | 5, 6 | 742 | |||
AK168003.1 | - | 5, 6 | 761 | |||
AK184796.1 | - | 1, 2 | 96 | |||
BC026983.1 | - | 5, 6 | 729 | 5365862 |