Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138595.2 | NP_613061.1 | 4 | 719 | ||
GenBank mRNA | AK080789.1 | - | 4 | 691 | ||
AK080937.1 | - | 4 | 708 | |||
AK139929.1 | - | 4 | 710 | |||
BC017135.1 | - | 4 | 696 | 4511573 |