Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138597.2 | NP_613063.1 | 5 | 449 | ||
GenBank mRNA | AK005309.1 | - | 5 | 448 | ||
AK168292.1 | - | 5 | 404 | |||
AK169318.1 | - | 5 | 383 | |||
BC012241.1 | - | 5 | 397 | 4023318 | ||
BC096643.1 | - | 5 | 407 |