Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138646.3 | NP_619587.3 | 5 | 861 | ||
GenBank mRNA | AK081489.1 | - | 5 | 821 | ||
AK129418.1 | - | 4 | 748 | |||
AK147954.1 | - | 4 | 1304 | |||
AK171871.1 | - | 5 | 830 | |||
AK187573.1 | - | 2 | 94 | |||
AY043415.1 | - | 4 | 381 | |||
BC063055.1 | - | 4 | 553 | 5701206 |